Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Diiodofluorene 98.0+%, TCI America™
SDP

Catalog No. D45041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45041G 1 g
D45045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45041G Supplier TCI America Supplier No. D45041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16218-28-3
Molecular Formula C13H8I2
Molecular Weight (g/mol) 418.016
MDL Number MFCD00040153
InChI Key YCWGCTPMBCOCLT-UHFFFAOYSA-N
PubChem CID 634493
IUPAC Name 2,7-diiodo-9H-fluorene
SMILES C1C2=C(C=CC(=C2)I)C3=C1C=C(C=C3)I

Specifications

Melting Point 218°C
Color Yellow
Quantity 1 g
Formula Weight 418.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diiodofluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.