Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Diisopropylnaphthalene 97.0+%, TCI America™
SDP

Catalog No. D242225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D242225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D242225ML Supplier TCI America Supplier No. D242225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40458-98-8
Molecular Formula C16H20
Molecular Weight (g/mol) 212.34
MDL Number MFCD00671522
InChI Key YGDMAJYAQCDTNG-UHFFFAOYSA-N
PubChem CID 94505
IUPAC Name 2,7-bis(propan-2-yl)naphthalene
SMILES CC(C)C1=CC2=CC(=CC=C2C=C1)C(C)C

Specifications

Color Yellow
Boiling Point 280°C
Quantity 25 mL
Formula Weight 212.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,7-Diisopropylnaphthalene
EINECSNumber (4)-0961
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.