Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,7-Dimethyl-9-Mesityl-10-Methylacridinium Tetrafluoroborate

Catalog No. 909181250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-918-1250MG 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-918-1250MG Supplier Sigma Aldrich Supplier No. 909181250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Robust acridinium-based photocatalyst reported by Nicewicz and coworkers as an alternative to transition-metal-based photocatalysts with higher chemical stability and attenuated redox potential.The direct anti-Markovnikov addition of mineral acids to styrenes

Specifications

CAS 1286281-32-0
Molecular Formula Pt
Quantity 10 g
Synonym Pt/C
Molecular Weight (g/mol) 195.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.