Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dinitrofluorene 97.0+%, TCI America™
SDP

Catalog No. D08345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08345G 5 g
D083425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08345G Supplier TCI America Supplier No. D08345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5405-53-8
Molecular Formula C13H8N2O4
Molecular Weight (g/mol) 256.217
MDL Number MFCD00001121
InChI Key IHZCVUBSTYOFSJ-UHFFFAOYSA-N
PubChem CID 21502
IUPAC Name 2,7-dinitro-9H-fluorene
SMILES C1C2=C(C=CC(=C2)[N+](=O)[O-])C3=C1C=C(C=C3)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 5 g
Synonym 2,7-dinitrofluorene, 9h-fluorene, 2,7-dinitro, fluorene, 2,7-dinitro, ccris 2909, fluorene,7-dinitro, acmc-1cuci, 9h-fluorene,7-dinitro, 9h-fluorene,2,7-dinitro, 4-05-00-02153 beilstein handbook reference
Formula Weight 256.22
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dinitrofluorene
RTECSNumber LL7175000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.