Learn More
BioSpec Products 2.7 MM GLASS BEADS, 1 LB
SDP
Catalog No. NC0911353
Encompass_Preferred
Glass Beads - 2.7mm dia. Glass has a density of 2.5 g/cc and is the most commonly used bead media for 'Beadbeating'. Beads are sold in one pound bottles.
Catalog No. NC0911353 Supplier BioSpec Products Supplier No. 11079127
Glass Beads - 2.7mm dia. Glass has a density of 2.5 g/cc and is the most commonly used bead media for 'Beadbeating'. Beads are sold in one pound bottles.