Learn More
Med Vet International 2.7 TPO/DPO Locking Plate 20 Left, J1212L
Catalog No. 5000581472
Encompass_Preferred
Triple Pelvic Osteotomy (TPO) and Double Pelvic Osteotomy (DPO) remain effrective techniques for the managment of hip dysplasia. Use of diverging, locking screws increases plate security as does the sightly larger footprint of our plates. Available in both 2.7 and 3.5 locking screw sizes at the following angles: 20.25 and 30 degrees. Extra long locking drill guides are available. 2.7mm TPO/DPO Locking Plate 20 Left.
Catalog No. 50-005-81472 Supplier Med Vet International Supplier No. JOR1212L
Triple Pelvic Osteotomy (TPO) and Double Pelvic Osteotomy (DPO) remain effrective techniques for the managment of hip dysplasia. Use of diverging, locking screws increases plate security as does the sightly larger footprint of our plates. Available in both 2.7 and 3.5 locking screw sizes at the following angles: 20.25 and 30 degrees. Extra long locking drill guides are available. 2.7mm TPO/DPO Locking Plate 20 Left.