Learn More
Perkin Elmer US LLC 2.79 mm I.D. Solvent Flex Tubing, Qty. 10 ft
Catalog No. 502117140
Encompass_Preferred
This tubing, made of solvent flex (yellow tint) PVC, is recommended for use with oils and many organic solvents.
Catalog No. 50-211-7140 Supplier Perkin Elmer US LLC Supplier No. 09985735
This tubing, made of solvent flex (yellow tint) PVC, is recommended for use with oils and many organic solvents.