Learn More
Cayman Chemical 2,7-Dichlorodihydrofluorescein diacetate, Purity: ≥95%, CAS: 4091-99-0, 50 mg
SDP
Catalog No. 502894800
Encompass_Preferred
A fluorescent indicator of peroxynitrite formation; neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DCFH
Catalog No. 50-289-4800 Supplier Cayman Chemical Supplier No. 8515550 MG
A fluorescent indicator of peroxynitrite formation; neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DCFH