Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,8-Dibromodibenzothiophene 96.0+%, TCI America™
SDP

Catalog No. D22451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22451G 1 g
D22455G 5 g
D224525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22451G Supplier TCI America Supplier No. D22451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31574-87-5
Molecular Formula C12H6Br2S
Molecular Weight (g/mol) 342.05
MDL Number MFCD00092755
InChI Key WNEXSUAHKVAPFK-UHFFFAOYSA-N
PubChem CID 631120
IUPAC Name 4,12-dibromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
SMILES BrC1=CC2=C(SC3=C2C=C(Br)C=C3)C=C1

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 1 g
Formula Weight 342.05
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Dibromodibenzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.