Learn More
Medchemexpress LLC 2,8-Quinolinediol | 15450-76-7 | 99.7% | 161.16 | 25 G
SDP
Catalog No. 5000403027
Encompass_Preferred
8-Hydroxy-2(1H)-quinolinone is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. This product is for research use only and has not been fully validated for medical applications.
  • Suitable for life science-related research
  • Can be used as a biological material or organic compound
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 20 mg/mL (124.10 mM) with ultrasonic, warming, and heating to 60°C
  • Forms clear solutions (≥ 2 mg/mL) in common in vivo dissolution protocols
Catalog No. 50-004-03027 Supplier Medchemexpress LLC Supplier No. 25GHY34494
8-Hydroxy-2(1H)-quinolinone is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. This product is for research use only and has not been fully validated for medical applications.
  • Suitable for life science-related research
  • Can be used as a biological material or organic compound
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 20 mg/mL (124.10 mM) with ultrasonic, warming, and heating to 60°C
  • Forms clear solutions (≥ 2 mg/mL) in common in vivo dissolution protocols