Learn More
Felix Storch Inc ACCUCOLD 2.83 Cubic Feet Vaccine Refrigerator ADA HT
SDP
Catalog No. NC2111140
Encompass_Preferred
The ARS32PVBIADA is a 2.83 cu.ft. all-refrigerator with a temperature range of +2°C to +8°C. Sized at 32" high to fit under lower counters complying with ADA guidelines for accessibility. designed and purpose-built for pharmacy, medication, and vaccine applications to support meeting CDC/VFC/VFA guidelines: https://www.accucold.com/product/ARS32PVBIADA
This item is not returnable. View return policy
Catalog No. NC2111140 Supplier Felix Storch Inc Supplier No. ARS32PVBIADA
The ARS32PVBIADA is a 2.83 cu.ft. all-refrigerator with a temperature range of +2°C to +8°C. Sized at 32" high to fit under lower counters complying with ADA guidelines for accessibility. designed and purpose-built for pharmacy, medication, and vaccine applications to support meeting CDC/VFC/VFA guidelines: https://www.accucold.com/product/ARS32PVBIADA