Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(9,9'-Spirobi[fluoren]-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
SDP

Catalog No. S09801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09801G 1 g
S09805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09801G Supplier TCI America Supplier No. S09801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 884336-44-1
Molecular Formula C31H27BO2
Molecular Weight (g/mol) 442.365
InChI Key VZMLUIPYIYNRGP-UHFFFAOYSA-N
PubChem CID 60135725
IUPAC Name 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-2-yl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C35C6=CC=CC=C6C7=CC=CC=C57

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 1 g
Synonym (9,9′C-Spirobi[fluoren]-7-yl)boronic Acid Pinacol Ester, 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′C-spirobi[fluorene]
Formula Weight 442.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9,9′-Spirobi[fluoren]-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.