Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,9-Dibutyl-1,10-phenanthroline 98.0+%, TCI America™
SDP

Catalog No. D2565100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2565100MG 100 mg
D25651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2565100MG Supplier TCI America Supplier No. D2565100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85575-93-5
Molecular Formula C20H24N2
Molecular Weight (g/mol) 292.426
MDL Number MFCD02093459
InChI Key LSGGPELKXXFMGO-UHFFFAOYSA-N
PubChem CID 13384176
IUPAC Name 2,9-dibutyl-1,10-phenanthroline
SMILES CCCCC1=NC2=C(C=CC3=C2N=C(C=C3)CCCC)C=C1

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 100 mg
Formula Weight 292.43
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,9-Dibutyl-1,10-phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.