Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,9-Dimethyl-5-picrylamino-1,10-phenanthroline, TCI America™
SDP

Catalog No. D2583100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2583100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2583100MG Supplier TCI America Supplier No. D2583100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 380482-30-4
Molecular Formula C20H14N6O6
Molecular Weight (g/mol) 434.368
MDL Number MFCD01861283
InChI Key YMWCRDXLDDCILP-UHFFFAOYSA-N
Synonym 2,9-Dimethyl-5-(2,4,6-trinitroanilino)-1,10-phenanthroline
PubChem CID 5068020
IUPAC Name 2,9-dimethyl-N-(2,4,6-trinitrophenyl)-1,10-phenanthrolin-5-amine
SMILES CC1=NC2=C3C(=C(C=C2C=C1)NC4=C(C=C(C=C4[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-])C=CC(=N3)C

Specifications

Quantity 100 mg
UN Number 1325
Formula Weight 434.37
Physical Form Crystalline Powder
Chemical Name or Material 2,9-Dimethyl-5-picrylamino-1,10-phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.