Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(9-Oxoxanthen-2-yl)propionic Acid 1,5-Diazabicyclo[4.3.0]non-5-ene Salt 98.0+%, TCI America™
SDP

Catalog No. O04471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04471G Supplier TCI America Supplier No. O04471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1346753-04-5
Molecular Formula C23H24N2O4
Molecular Weight (g/mol) 392.455
InChI Key OQEMAQQKYSJWQK-UHFFFAOYSA-N
PubChem CID 134128833
IUPAC Name 2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine;2-(9-oxoxanthen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)OC3=CC=CC=C3C2=O)C(=O)O.C1CC2=NCCCN2C1

Specifications

Color White
Quantity 1 g
Formula Weight 392.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9-Oxoxanthen-2-yl)propionic Acid 1,5-Diazabicyclo[4.3.0]non-5-ene Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.