Learn More
Eurofins Discoverx PathHunter Bioassay Detection Kit, 15 (200 dp, 96-well). (93-0933E)
Catalog No. NC1318472
Encompass
The PathHunter Bioassay Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter Bioassay cells or cell lines. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 2 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0933e.
Catalog No. NC1318472 Supplier Eurofins Discoverx Supplier No. 930933E
The PathHunter Bioassay Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter Bioassay cells or cell lines. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 2 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0933e.