Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-AAPA hydrate 95 HPLC5MG
Catalog No. 502901661
Encompass
2-Acetylamino-3-[4-(2-acetylamino-2-carboxy-ethylsulfanylthiocarbonylamino)phenylthiocarbamoylsulfanyl]propionic acid (2-AAPA) can block reductase systems thereby modulating peroxiredoxin oxidation and mitochondrial function in A172 glioblastoma cells.
Catalog No. 50-290-1661 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A41115MG
2-Acetylamino-3-[4-(2-acetylamino-2-carboxy-ethylsulfanylthiocarbonylamino)phenylthiocarbamoylsulfanyl]propionic acid (2-AAPA) can block reductase systems thereby modulating peroxiredoxin oxidation and mitochondrial function in A172 glioblastoma cells.