Learn More
Medchemexpress LLC 2-Acetamido-2 4-dide 1mg | 129728-87-6 | 223.20 g·mol⁻¹ | C8H14FNO5 | 1 MG
SDP
Catalog No. 5000308208
Encompass_Preferred
2-Acetamido-2,4-dideoxy-4-fluoro-α-D-glucopyranose is a synthetic monosaccharide derivative used in glycobiology research as a hepatic glycosaminoglycan biosynthesis inhibitor. It is provided as a high-purity solid for biochemical and cell-based assays.
  • High purity suitable for research applications.
  • Molecular weight 223.20 g·mol⁻¹ and formula C8H14FNO5.
  • Reported mechanism: inhibits hepatic glycosaminoglycan biosynthesis.
  • Supplied as a small solid quantity for assay development.
  • Intended for research use only; follow appropriate laboratory safety precautions.
Catalog No. 50-003-08208 Supplier Medchemexpress LLC Supplier No. HY1157381MG
2-Acetamido-2,4-dideoxy-4-fluoro-α-D-glucopyranose is a synthetic monosaccharide derivative used in glycobiology research as a hepatic glycosaminoglycan biosynthesis inhibitor. It is provided as a high-purity solid for biochemical and cell-based assays.
  • High purity suitable for research applications.
  • Molecular weight 223.20 g·mol⁻¹ and formula C8H14FNO5.
  • Reported mechanism: inhibits hepatic glycosaminoglycan biosynthesis.
  • Supplied as a small solid quantity for assay development.
  • Intended for research use only; follow appropriate laboratory safety precautions.