Learn More
Medchemexpress LLC 2-acetamido-2-deoxy-D-glucono-1,5-lactone | 19026-22-3 | MFCD00076154 | 90.0% | 219.19 g·mol⁻¹ | C8H13NO6 | 5 MG
SDP
Catalog No. 5000281059
Encompass_Preferred
2-Acetamido-2-deoxy-D-glucono-1,5-lactone (90%) is an N-acetylglucosamine 1,5-lactone supplied as a solid reagent for carbohydrate chemistry and biochemical assays. It is commonly used as a mechanistic inhibitor in glycosidase studies, and as an analytical reference for method development.
Catalog No. 50-002-81059 Supplier Medchemexpress LLC Supplier No. HYW4087895MG
2-Acetamido-2-deoxy-D-glucono-1,5-lactone (90%) is an N-acetylglucosamine 1,5-lactone supplied as a solid reagent for carbohydrate chemistry and biochemical assays. It is commonly used as a mechanistic inhibitor in glycosidase studies, and as an analytical reference for method development.