Learn More
Medchemexpress LLC 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride | 41355-44-6 | MFCD00047513 | >95.0% | 365.76 g/mol | C14H20ClNO8 | 1 G
SDP
Catalog No. 5000300220
Encompass_Preferred
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is a protected glycosyl chloride used as a glycosyl donor in glycochemistry and glycobiology research. It is commonly employed in the synthesis of oligosaccharides and glycan derivatives for analytical and preparative applications.
  • Protected glycosyl donor for oligosaccharide synthesis.
  • Facilitates regio- and stereoselective glycosylation reactions.
  • Reported molecular formula C14H20ClNO8 and molecular weight 365.76 g/mol.
  • Typical purity ≥95% suitable for research applications.
  • Available in small research quantities for method development and synthesis.
Catalog No. 50-003-00220 Supplier Medchemexpress LLC Supplier No. HYW0399281G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is a protected glycosyl chloride used as a glycosyl donor in glycochemistry and glycobiology research. It is commonly employed in the synthesis of oligosaccharides and glycan derivatives for analytical and preparative applications.
  • Protected glycosyl donor for oligosaccharide synthesis.
  • Facilitates regio- and stereoselective glycosylation reactions.
  • Reported molecular formula C14H20ClNO8 and molecular weight 365.76 g/mol.
  • Typical purity ≥95% suitable for research applications.
  • Available in small research quantities for method development and synthesis.