Learn More
Medchemexpress LLC 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride | 41355-44-6 | MFCD00047513 | 97.0% | 365.76 g/mol | C14H20ClNO8 | 5 G
SDP
Catalog No. 5000299784
Encompass_Preferred
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is an acetyl-protected amino-sugar glycosyl chloride used as a glycosyl donor in carbohydrate chemistry and glycobiology research. It is employed as an intermediate in the synthesis of protected glycosides and for method development in glycosylation and analytical workflows.
  • Acetyl-protected glycosyl donor suitable for glycosylation reactions.
  • Contains 2-acetamido functionality for amino-sugar derivatization.
  • Stable protected form for storage and handling in synthetic workflows.
  • Compatible with common glycosylation promoters and catalysts.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-99784 Supplier Medchemexpress LLC Supplier No. HYW0399285G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is an acetyl-protected amino-sugar glycosyl chloride used as a glycosyl donor in carbohydrate chemistry and glycobiology research. It is employed as an intermediate in the synthesis of protected glycosides and for method development in glycosylation and analytical workflows.
  • Acetyl-protected glycosyl donor suitable for glycosylation reactions.
  • Contains 2-acetamido functionality for amino-sugar derivatization.
  • Stable protected form for storage and handling in synthetic workflows.
  • Compatible with common glycosylation promoters and catalysts.
  • Available in small laboratory pack sizes for research use.