Learn More
Medchemexpress LLC 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride | 41355-44-6 | MFCD00047513 | >98.0% | 365.76 g/mol | C14H20ClNO8 | 500 MG
SDP
Catalog No. 5000299470
Encompass_Preferred
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is an acetyl-protected glycosyl chloride used as a glycosyl donor and synthetic intermediate in carbohydrate chemistry and glycobiology research. It is employed to assemble oligosaccharides and glycan analogues via glycosylation reactions and is handled under standard laboratory safety procedures.
  • Acetyl-protected glycosyl donor suitable for glycosylation reactions.
  • Facilitates synthesis of oligosaccharides and glycan analogues.
  • Reactive anomeric chloride enables coupling under standard activation conditions.
  • Compatible with common protecting group strategies in carbohydrate synthesis.
  • Supplied for research use; handle with appropriate PPE and protocols.
Catalog No. 50-002-99470 Supplier Medchemexpress LLC Supplier No. HYW039928500MG
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is an acetyl-protected glycosyl chloride used as a glycosyl donor and synthetic intermediate in carbohydrate chemistry and glycobiology research. It is employed to assemble oligosaccharides and glycan analogues via glycosylation reactions and is handled under standard laboratory safety procedures.
  • Acetyl-protected glycosyl donor suitable for glycosylation reactions.
  • Facilitates synthesis of oligosaccharides and glycan analogues.
  • Reactive anomeric chloride enables coupling under standard activation conditions.
  • Compatible with common protecting group strategies in carbohydrate synthesis.
  • Supplied for research use; handle with appropriate PPE and protocols.