Learn More
Medchemexpress LLC 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride | 41355-44-6 | MFCD00192160 | 97.0% | 365.76 g/mol | C14H20ClNO8 | 250 MG
SDP
Catalog No. 5000303513
Encompass_Preferred
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is an acetylated glycosyl chloride supplied as a powder for research use. It is used as an intermediate and building block in glycochemistry and glycobiology, particularly for glycosylation reactions and oligosaccharide synthesis.
  • Acetylated glycosyl chloride suitable for glycosylation reactions.
  • Used as a building block for oligosaccharide synthesis.
  • High purity (≈97%).
  • Powder form with recommended storage at -20°C or 4°C to preserve stability.
  • Molecular weight 365.76 g/mol.
Catalog No. 50-003-03513 Supplier Medchemexpress LLC Supplier No. 250MGHYW039928
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-galactopyranosyl chloride is an acetylated glycosyl chloride supplied as a powder for research use. It is used as an intermediate and building block in glycochemistry and glycobiology, particularly for glycosylation reactions and oligosaccharide synthesis.
  • Acetylated glycosyl chloride suitable for glycosylation reactions.
  • Used as a building block for oligosaccharide synthesis.
  • High purity (≈97%).
  • Powder form with recommended storage at -20°C or 4°C to preserve stability.
  • Molecular weight 365.76 g/mol.