Learn More
Medchemexpress LLC 2-acetamido-3-chloro-1,4-naphthoquinone | 5397-78-4 | 99.6% | 249.65 g/mol | C12H8ClNO3 | 5 MG
SDP
Catalog No. 5000263154
Encompass_Preferred
NP-313 is an antithrombotic small-molecule research compound that inhibits platelet aggregation and thromboxane A2 synthesis. Supplied as a light yellow solid with high analytical purity and predictable solubility in DMSO, it is intended for research use in studies of platelet function and thrombotic signaling pathways. Store the powder frozen for long-term stability.
Catalog No. 50-002-63154 Supplier Medchemexpress LLC Supplier No. HY1294965MG
NP-313 is an antithrombotic small-molecule research compound that inhibits platelet aggregation and thromboxane A2 synthesis. Supplied as a light yellow solid with high analytical purity and predictable solubility in DMSO, it is intended for research use in studies of platelet function and thrombotic signaling pathways. Store the powder frozen for long-term stability.