Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-acetamido-3-phenylprop-2-enoic acid | 5469-45-4 | MFCD00188416 | 99.5% | 205.21 g·mol⁻¹ | C11H11NO3 | 25 G
SDP

Catalog No. 5000303524
Encompass_Preferred

α-Acetamidocinnamic acid is an alkaloid-derived research reagent (CAS 5469-45-4) used in biochemical and natural product studies. It is supplied as a high-purity solid for use as a reference compound, synthetic building block, or analytical standard. Certificate of analysis and safety data sheet are available.

  • CAS number 5469-45-4.
  • Molecular formula C11H11NO3.
  • Molecular weight 205.21 g·mol⁻¹.
  • Purity 99.5%.
  • Supplied as a solid.
  • Available pack sizes: 25 g, 50 g, 100 g.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-003-03524 Supplier Medchemexpress LLC Supplier No. 25GHYW037875
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Acetamidocinnamic acid is an alkaloid-derived research reagent (CAS 5469-45-4) used in biochemical and natural product studies. It is supplied as a high-purity solid for use as a reference compound, synthetic building block, or analytical standard. Certificate of analysis and safety data sheet are available.

  • CAS number 5469-45-4.
  • Molecular formula C11H11NO3.
  • Molecular weight 205.21 g·mol⁻¹.
  • Purity 99.5%.
  • Supplied as a solid.
  • Available pack sizes: 25 g, 50 g, 100 g.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.