Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamido-5-bromo-4-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. A28231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28231G Supplier TCI America Supplier No. A28231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142404-82-8
Molecular Formula C8H9BrN2O
Molecular Weight (g/mol) 229.077
MDL Number MFCD06798092
InChI Key WVJCOPBDWMWUNP-UHFFFAOYSA-N
Synonym 2-Acetamido-5-bromo-4-picoline
PubChem CID 15179563
IUPAC Name N-(5-bromo-4-methylpyridin-2-yl)acetamide
SMILES CC1=CC(=NC=C1Br)NC(=O)C

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 1 g
Formula Weight 229.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-5-bromo-4-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.