Learn More
AdipoGen 2-Acetamido-5-mercapto-thiadia
SDP
Supplier:  AdipoGen CDXA0756G025
Encompass_Preferred
Chemical. CAS 32873-56-6. Formula C4H5N3OS2. MW 175.24. 2-Acetamido-5-mercapto-1,3,4-thiadiazole coatings are designed to prevent corrosion of metal ions by forming a barrier on metallic surfaces. 2-Acetylamino-5-mercapto-1,3,4-thiadiazole has been used in coatings as corrosion inhibitor for zinc and copper. 2-Acetamido-5-mercapto-1,3,4-thiadiazole is a intermediate and impurity acetazolamide carbonic anhydrase inhibitor and has been used for synthesis in organic chemistry.
Catalog No. 50-256-1719