Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamido-5-nitrothiazole 98.0+%, TCI America™
SDP

Catalog No. A06895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06895G 5 g
A06891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06895G Supplier TCI America Supplier No. A06895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 140-40-9
Molecular Formula C5H5N3O3S
Molecular Weight (g/mol) 187.17
MDL Number MFCD00022438
InChI Key UJRRDDHEMZLWFI-UHFFFAOYSA-N
Synonym nithiamide, 2-acetamido-5-nitrothiazole, aminitrozole, acinitrazole, nitazole, tritheon, gynofon, ametoterina, aminitrozol, lavoflagin
PubChem CID 8798
IUPAC Name N-(5-nitro-1,3-thiazol-2-yl)acetamide
SMILES CC(=O)NC1=NC=C(S1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 187.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-5-nitrothiazole
EINECSNumber (9)-2179&(9)-1849
RTECSNumber XJ1570000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.