Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Acetamidofluorene | 53-96-3 | 223.28 | 1 ML
SDP

Catalog No. 5000454309
Encompass_Preferred

2-Acetamidofluorene is a potent carcinogen primarily used for the induction of hepatocellular carcinoma (HCC) and multiple primary tumors. It demonstrates strong damaging effects on DNA and is commonly employed in studies involving hepatic cell degeneration, oval cell proliferation, and inflammatory cell infiltration in rat liver tissues.

  • Potent carcinogen.
  • Used for induction of hepatocellular carcinoma (HCC) and multiple primary tumors.
  • Exhibits strong damaging effects on DNA.
  • Targets DNA/RNA synthesis.
  • Involves cell cycle/DNA damage pathway.
  • Storage at 4°C, protected from light.
  • Soluble in DMSO at 125 mg/mL.

Catalog No. 50-004-54309 Supplier Medchemexpress LLC Supplier No. HYW01351410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Acetamidofluorene is a potent carcinogen primarily used for the induction of hepatocellular carcinoma (HCC) and multiple primary tumors. It demonstrates strong damaging effects on DNA and is commonly employed in studies involving hepatic cell degeneration, oval cell proliferation, and inflammatory cell infiltration in rat liver tissues.

  • Potent carcinogen.
  • Used for induction of hepatocellular carcinoma (HCC) and multiple primary tumors.
  • Exhibits strong damaging effects on DNA.
  • Targets DNA/RNA synthesis.
  • Involves cell cycle/DNA damage pathway.
  • Storage at 4°C, protected from light.
  • Soluble in DMSO at 125 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.