Learn More
Medchemexpress LLC 2-Acetamidofluorene | 53-96-3 | 223.28 | 250 MG
SDP
Catalog No. 5000367047
Encompass_Preferred
2-Acetamidofluorene is a potent carcinogen and can be used for the induction of hepatocellular carcinoma (HCC) and multiple primary tumors. It exhibits strong damaging effects on DNA.
  • Potent carcinogen
  • Used for induction of hepatocellular carcinoma (HCC) and multiple primary tumors
  • Exhibits strong damaging effects on DNA
  • Used to induce hepatocellular carcinoma (HCC) Model
  • Induction of hepatocellular carcinoma (HCC) model
Catalog No. 50-003-67047 Supplier Medchemexpress LLC Supplier No. 250MGHYW013514
2-Acetamidofluorene is a potent carcinogen and can be used for the induction of hepatocellular carcinoma (HCC) and multiple primary tumors. It exhibits strong damaging effects on DNA.
  • Potent carcinogen
  • Used for induction of hepatocellular carcinoma (HCC) and multiple primary tumors
  • Exhibits strong damaging effects on DNA
  • Used to induce hepatocellular carcinoma (HCC) Model
  • Induction of hepatocellular carcinoma (HCC) model