Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-acetamidophenol (orthocetamol) | 614-80-2 | MFCD00002181 | 99.6% | 151.16 | C8H9NO2 | 10 G
SDP

Catalog No. 5000300066
Encompass_Preferred

2-Acetamidophenol (orthocetamol) is the ortho-regioisomer of paracetamol and is used as a research reagent that modulates ferroptosis and glutathione metabolic pathways. It has been reported to reduce intracellular reactive oxygen species and ferrous ion accumulation, enhance glutathione peroxidase activity, and influence lipid levels in model systems.

  • Regulates ferroptosis and glutathione metabolism.
  • Reduces intracellular ROS and Fe2+ accumulation.
  • Enhances GPX4 activity and supports glutathione synthesis gene expression.
  • High stated purity (99.61%).
  • Molecular weight 151.16; formula C8H9NO2.
  • CAS number 614-80-2 for unambiguous identification.

Catalog No. 50-003-00066 Supplier Medchemexpress LLC Supplier No. HYW01560010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Acetamidophenol (orthocetamol) is the ortho-regioisomer of paracetamol and is used as a research reagent that modulates ferroptosis and glutathione metabolic pathways. It has been reported to reduce intracellular reactive oxygen species and ferrous ion accumulation, enhance glutathione peroxidase activity, and influence lipid levels in model systems.

  • Regulates ferroptosis and glutathione metabolism.
  • Reduces intracellular ROS and Fe2+ accumulation.
  • Enhances GPX4 activity and supports glutathione synthesis gene expression.
  • High stated purity (99.61%).
  • Molecular weight 151.16; formula C8H9NO2.
  • CAS number 614-80-2 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.