Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-acetamidophenol (orthocetamol) | 614-80-2 | MFCD00002181 | 99.6% | 151.16 g/mol | C8H9NO2 | 100 G
SDP

Catalog No. 5000300401
Encompass_Preferred

2-Acetamidophenol (orthocetamol) is the ortho-regioisomer of acetaminophen provided as a research reagent for biochemical and pharmacological studies, including investigations of ferroptosis and glutathione metabolism. The compound is supplied as a solid with standard analytical characterization and is intended for laboratory use under appropriate safety controls.

  • High purity suitable for research applications.
  • Molecular formula C8H9NO2.
  • Molecular weight 151.16 g/mol.
  • Supplied as a solid in common laboratory pack sizes.
  • Applicable for biochemical and cellular assay research.
  • Handle according to the safety data sheet and laboratory safety practices.

Catalog No. 50-003-00401 Supplier Medchemexpress LLC Supplier No. HYW015600100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Acetamidophenol (orthocetamol) is the ortho-regioisomer of acetaminophen provided as a research reagent for biochemical and pharmacological studies, including investigations of ferroptosis and glutathione metabolism. The compound is supplied as a solid with standard analytical characterization and is intended for laboratory use under appropriate safety controls.

  • High purity suitable for research applications.
  • Molecular formula C8H9NO2.
  • Molecular weight 151.16 g/mol.
  • Supplied as a solid in common laboratory pack sizes.
  • Applicable for biochemical and cellular assay research.
  • Handle according to the safety data sheet and laboratory safety practices.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.