Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences 2% Acetocarmine, Aqueous
SCI_ED

Catalog No. S000786
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000786 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000786 Supplier G-Biosciences Supplier No. 7862202
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2% Acetocarmine, Aqueous solution is used to visualize nucleic acids, DNA and RNA.

This stain solution is used for cytogenetic studies and chromosome analysis.

Specifications

Quantity 250 mL
Product Type Acetocarmine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.