Learn More
Medchemexpress LLC 2-acetonaphthone | 93-08-3 | MFCD00004108 | 1mL
SDP
Supplier:  Medchemexpress LLC HYY181910MM/1ML
Encompass_Preferred
2-Acetonaphthone is a synthetic fragrance material 2-Acetonaphthone increases ROS under UVA/sunlight leading to endoplasmic reticulum stress and decreased mitochondrial membrane potential 2-Acetonaphthone can be used as an adulterant in a variety of cosmetics 2-Acetonaphthone can be used for the study of skin keratinization[1]
Catalog No. 50-002-66176