Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-[(Acetoxymethyl)allyl]trimethylsilane

Catalog No. 3398305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-983-05G 5 g
33-983-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-983-05G Supplier Sigma Aldrich Supplier No. 3398305G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-[(Acetoxymethyl)allyl]trimethylsilane was used in the cyclopentane synthesis via Pd(0)-catalyzed reactions with 2-acceptors.

Specifications

CAS 72047-94-0
Density 0.877 g/mL (at 25°C (literature))
Boiling Point 95°C (7 mmHg)
Molecular Formula C9H18O2Si
Refractive Index n20/D 1.440 (literature)
Linear Formula CH3CO2CH2C(=CH2)CH2Si(CH3)3
Quantity 5 g
MDL Number MFCD00075170
Synonym 2-(Trimethylsilylmethyl)-2-propen-1-yl acetate; 2-(Trimethylsilylmethyl)allyl acetate; 2-Acetoxymethyl-3-(trimethylsilyl)propene
Molecular Weight (g/mol) 186.32
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.