Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetoxypropionic Acid 98.0+%, TCI America™
SDP

Catalog No. A29315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29315G 5 g
A29311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29315G Supplier TCI America Supplier No. A29315G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 535-17-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00674491
InChI Key WTLNOANVTIKPEE-UHFFFAOYNA-N
Synonym O-Acetyllactic Acid
PubChem CID 79041
IUPAC Name 2-(acetyloxy)propanoic acid
SMILES CC(OC(C)=O)C(O)=O

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 g
Formula Weight 132.12
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Acetoxypropionic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.