Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-3-ethylpyrazine 98.0+%, TCI America™
SDP

Catalog No. A20035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20035G 5 g
A20031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20035G Supplier TCI America Supplier No. A20035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32974-92-8
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00038028
InChI Key PPJSYGVFDJEMRP-UHFFFAOYSA-N
Synonym 2-acetyl-3-ethylpyrazine, 1-3-ethylpyrazin-2-yl ethanone, ethanone, 1-3-ethylpyrazinyl, 2-acetyl-3-ethyl-1,4-diazine, 1-3-ethylpyrazinyl ethan-1-one, unii-aq03n14xtl, 1-3-ethylpyrazinyl ethanone, 1-3-ethyl-2-pyrazinyl ethanone, 3-ethyl-2-acetylpyrazine, 2-acetyl-3-ethyl pyrazine
PubChem CID 61918
IUPAC Name 1-(3-ethylpyrazin-2-yl)ethanone
SMILES CCC1=NC=CN=C1C(=O)C

Specifications

Color Yellow
Boiling Point 77°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetyl-3-ethylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.