Learn More
eMolecules​ 2-Acetyl-3-thienylboronic acid | 36155-74-5 | 5G | Purity: 95%
Supplier:  eMolecules​ 117526393
Encompass_Preferred
Combi-Blocks | 2-Acetyl-3-thienylboronic acid | 5G | 36155-74-5 | MFCD01318932
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-158-3548