Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-4-bromopyridine, 98%, Thermo Scientific™

Catalog No. AAH66562MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66562MD 250 mg
AAH6656203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66562MD Supplier Thermo Scientific Chemicals Supplier No. H66562MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1060805-69-7
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.04
MDL Number MFCD13189212
InChI Key NVVSJVCWQHRSKT-UHFFFAOYSA-N
Synonym 1-4-bromopyridin-2-yl ethanone, 1-4-bromopyridin-2-yl ethan-1-one, 1-4-bromo-2-pyridinyl ethanone, 1-4-bromanylpyridin-2-yl ethanone, ethanone, 1-4-bromo-2-pyridinyl, 1-4-bromo-pyridin-2-yl-ethanone
PubChem CID 23437052
IUPAC Name 1-(4-bromopyridin-2-yl)ethan-1-one
SMILES CC(=O)C1=NC=CC(Br)=C1

Specifications

Quantity 250 mg
Beilstein 22133194
Formula Weight 200.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Acetyl-4-bromopyridine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.