Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-4-methylthiophene, 98%, Thermo Scientific™

Catalog No. AAL1761706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1761706 5 g
AAL1761703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1761706 Supplier Thermo Scientific Chemicals Supplier No. L1761706

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13679-73-7
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00027395
InChI Key NFZNHUNPIJPRMP-UHFFFAOYSA-N
Synonym 2-acetyl-4-methylthiophene, 1-4-methylthiophen-2-yl ethanone, 1-4-methyl-2-thienyl ethanone, ethanone, 1-4-methyl-2-thienyl, 1-4-methyl-2-thienyl ethan-1-one, 1-4-methyl-thiophen-2-yl-ethanone, 4-methyl-2-acetylthiophene, pubchem5214, acmc-20aa2e
PubChem CID 83654
IUPAC Name 1-(4-methylthiophen-2-yl)ethanone
SMILES CC1=CSC(=C1)C(=O)C

Specifications

Boiling Point 97°C to 99°C (8 mmHg)
Refractive Index 1.56
Quantity 5 g
Beilstein 111378
Formula Weight 140.2
Percent Purity 98%
Chemical Name or Material 2-Acetyl-4-methylthiophene
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 237-180-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.