Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-6-(pentafluoroethyl)pyridine, 96%, Thermo Scientific™

Catalog No. AAH66942MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66942MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66942MC Supplier Thermo Scientific Chemicals Supplier No. H66942MC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1580464-65-8
Molecular Formula C9H6F5NO
Molecular Weight (g/mol) 239.145
InChI Key KCNUAVPBVPTCLU-UHFFFAOYSA-N
PubChem CID 99728608
IUPAC Name 1-[6-(1,1,2,2,2-pentafluoroethyl)pyridin-2-yl]ethanone
SMILES CC(=O)C1=NC(=CC=C1)C(C(F)(F)F)(F)F

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 239.14
Percent Purity 96%
Physical Form Viscous Liquid
Chemical Name or Material 2-Acetyl-6-(pentafluoroethyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.