Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-7-methoxybenzofuran 98.0+%, TCI America™
SDP

Catalog No. A131710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A131710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A131710G Supplier TCI America Supplier No. A131710G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 43071-52-9
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00016911
InChI Key MUUMFANQVPWPFY-UHFFFAOYSA-N
Synonym 2-acetyl-7-methoxybenzofuran, 1-7-methoxybenzofuran-2-yl ethanone, ethanone, 1-7-methoxy-2-benzofuranyl, 1-7-methoxy-1-benzofuran-2-yl ethanone, 2-acetyl-7-methoxybenzo b furan, 2-acetyl-7-methoxycoumarone, 7-methoxy-2-benzofuranyl methyl ketone, ketone, 7-methoxy-2-benzofuranyl methyl, 1-7-methoxy-2-benzofuranyl ethanone, 1-7-methoxy-1-benzofuran-2-yl ethan-1-one
PubChem CID 39378
IUPAC Name 1-(7-methoxy-1-benzofuran-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(O1)C(=CC=C2)OC

Specifications

Melting Point 93°C
Color Yellow
Boiling Point 180°C
Quantity 10 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-7-methoxybenzofuran
RTECSNumber OB4571050
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.