Learn More
Medchemexpress LLC 2-(acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose | 55637-63-3 | 98.0% | 383.35 Da | C14H25NO11 | 5 MG
SDP
Catalog No. 5000284038
Encompass_Preferred
2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose is a glycosylated N-acetyl amino sugar used as a biochemical reagent in glycobiology and carbohydrate research. Supplied as a solid for laboratory assays and synthesis, it is used as a defined glycoside building block or analytical standard.
  • High purity suitable for analytical and synthetic applications (reported ≥98%).
  • Useful as a glycoside building block or standard in glycobiology assays.
  • Stable solid form; recommended storage at -20°C (powder) or -80°C in solution.
  • Provided in small sample sizes appropriate for research use.
Catalog No. 50-002-84038 Supplier Medchemexpress LLC Supplier No. HYW6990375MG
2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose is a glycosylated N-acetyl amino sugar used as a biochemical reagent in glycobiology and carbohydrate research. Supplied as a solid for laboratory assays and synthesis, it is used as a defined glycoside building block or analytical standard.
  • High purity suitable for analytical and synthetic applications (reported ≥98%).
  • Useful as a glycoside building block or standard in glycobiology assays.
  • Stable solid form; recommended storage at -20°C (powder) or -80°C in solution.
  • Provided in small sample sizes appropriate for research use.