Learn More
Medchemexpress LLC 2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose | 55637-63-3 | MFCD01310846 | 98.0% | 383.35 g·mol⁻¹ | C14H25NO11 | 10 MG
SDP
Catalog No. 5000284319
Encompass_Preferred
2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose is a glycosylated amino sugar reagent used in glycobiology and carbohydrate chemistry research. Supplied as a powder with high purity and good aqueous solubility, it is suitable for enzymology, glycan synthesis, and analytical applications.
  • Powder form, readily soluble in water (125 mg/mL with ultrasonic).
  • High purity (98.0%) suitable for research use.
  • Molecular weight 383.35 g·mol⁻¹ for accurate mass calculations.
  • Stable under frozen storage (powder: -20 °C, shelf life ~3 years).
  • Available in small research pack sizes including 10 MG for analytical experiments.
Catalog No. 50-002-84319 Supplier Medchemexpress LLC Supplier No. HYW69903710MG
2-(Acetylamino)-2-deoxy-4-O-β-D-mannopyranosyl-D-glucose is a glycosylated amino sugar reagent used in glycobiology and carbohydrate chemistry research. Supplied as a powder with high purity and good aqueous solubility, it is suitable for enzymology, glycan synthesis, and analytical applications.
  • Powder form, readily soluble in water (125 mg/mL with ultrasonic).
  • High purity (98.0%) suitable for research use.
  • Molecular weight 383.35 g·mol⁻¹ for accurate mass calculations.
  • Stable under frozen storage (powder: -20 °C, shelf life ~3 years).
  • Available in small research pack sizes including 10 MG for analytical experiments.