Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(acetylamino)-2-deoxy-β-D-glucopyranosylamine | 4229-38-3 | MFCD00057751 | 98.4% | 220.22 | C8H16N2O5 | 5 MG
SDP

Catalog No. 5000282146
Encompass_Preferred

2-(Acetylamino)-2-deoxy-β-D-glucopyranosylamine is a purified carbohydrate derivative used in glycobiology and biochemical research. Supplied as a solid in milligram quantities, it is characterized for identity and purity and provided with a product datasheet and safety data for laboratory handling.

  • Purity 98.36%.
  • Solid powder form for storage and handling.
  • Molecular weight 220.22 g/mol.
  • Chemical formula C8H16N2O5.
  • Storage: powder -20°C for long term, 4°C for shorter term; in solvent -80°C recommended.
  • Available in milligram quantities suitable for analytical and synthetic applications.

Catalog No. 50-002-82146 Supplier Medchemexpress LLC Supplier No. HYW3413015MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Acetylamino)-2-deoxy-β-D-glucopyranosylamine is a purified carbohydrate derivative used in glycobiology and biochemical research. Supplied as a solid in milligram quantities, it is characterized for identity and purity and provided with a product datasheet and safety data for laboratory handling.

  • Purity 98.36%.
  • Solid powder form for storage and handling.
  • Molecular weight 220.22 g/mol.
  • Chemical formula C8H16N2O5.
  • Storage: powder -20°C for long term, 4°C for shorter term; in solvent -80°C recommended.
  • Available in milligram quantities suitable for analytical and synthetic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.