Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Acetylamino)-2-deoxy-β-D-glucopyranosylamine | 4229-38-3 | MFCD00057751 | 98.4% | 220.22 g/mol | C8H16N2O5 | 10 MG
SDP

Catalog No. 5000282411
Encompass_Preferred

2-(Acetylamino)-2-deoxy-β-D-glucopyranosylamine is a white to off-white solid biochemical reagent used in glycobiology research and small-scale synthesis. It is provided in mg-scale quantities with supporting analytical documentation to enable characterization and reproducible use in laboratory assays.

  • Purity 98.36%
  • Cas number 4229-38-3
  • Molecular weight 220.22 g/mol
  • Molecular formula C8H16N2O5
  • Available in small mg-scale packages for analytical and synthetic work
  • Includes analytical documents: COA, HNMR, RP-HPLC, LCMS, and SDS
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-82411 Supplier Medchemexpress LLC Supplier No. HYW34130110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Acetylamino)-2-deoxy-β-D-glucopyranosylamine is a white to off-white solid biochemical reagent used in glycobiology research and small-scale synthesis. It is provided in mg-scale quantities with supporting analytical documentation to enable characterization and reproducible use in laboratory assays.

  • Purity 98.36%
  • Cas number 4229-38-3
  • Molecular weight 220.22 g/mol
  • Molecular formula C8H16N2O5
  • Available in small mg-scale packages for analytical and synthetic work
  • Includes analytical documents: COA, HNMR, RP-HPLC, LCMS, and SDS
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.