Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Acetylamino-3-(2-quinolon-4-yl)propionic acid | 681806-75-7 | 274.28 | 100 MG
SDP

Catalog No. 5000404531
Encompass_Preferred

2-Acetylamino-3-(2-quinolon-4-yl)propionic acid is a drug intermediate for synthesis of various active compounds. It is for research use only and not sold to patients.

  • Molecular weight: 274.28
  • Formula: C₁₄H₁₄N₂O₄
  • CAS No.: 681806-75-7
  • Appearance: Solid
  • Target: Drug intermediate
  • Storage conditions: -20°C for powder, -80°C in solvent
  • Solubility: DMSO: 50 mg/mL

Catalog No. 50-004-04531 Supplier Medchemexpress LLC Supplier No. HY41638100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Acetylamino-3-(2-quinolon-4-yl)propionic acid is a drug intermediate for synthesis of various active compounds. It is for research use only and not sold to patients.

  • Molecular weight: 274.28
  • Formula: C₁₄H₁₄N₂O₄
  • CAS No.: 681806-75-7
  • Appearance: Solid
  • Target: Drug intermediate
  • Storage conditions: -20°C for powder, -80°C in solvent
  • Solubility: DMSO: 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.