Learn More
Medchemexpress LLC 2-Acetylbenzoic acid | 577-56-0 | 164.16 | 100 G
SDP
Supplier:  Medchemexpress LLC HYN1717100G
Encompass_Preferred
2-Acetylbenzoic acid is a Benzoic acid derivative that acts as a weak platelet aggregation inhibitor. It specifically inhibits Adenosine 5'-diphosphate (ADP)-induced platelet aggregation. This product is intended for research use only and is not for human use.
  • Inhibits ADP-induced platelet aggregation
  • Suitable for various research applications
  • Available in solid form
Catalog No. 50-004-16012