Learn More
Medchemexpress LLC 2-Acetylbenzoic acid | 577-56-0 | 164.16 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN171750G
Encompass_Preferred
2-Acetylbenzoic acid is a derivative of Benzoic acid that functions as a weak platelet aggregation inhibitor. It inhibits Adenosine 5'-diphosphate (ADP)-induced platelet aggregation. This product is intended for research use only.
  • Inhibits ADP-induced platelet aggregation
  • Available in multiple quantities
  • Off-white to yellow solid appearance
  • Can be stored for 3 years at room temperature
Catalog No. 50-004-16538