Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylbenzoic acid, 99%

Catalog No. AC102471000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102471000 100 g Glass bottle
AC102470250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102471000 Supplier Thermo Scientific Chemicals Supplier No. 102471000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 577-56-0 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-56-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002475
InChI Key QDAWXRKTSATEOP-UHFFFAOYSA-N
Synonym o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
PubChem CID 68474
IUPAC Name 2-acetylbenzoic acid
SMILES CC(=O)C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 118°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3COC6H4COOH
Quantity 100 g
Beilstein 10, 690
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylbenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-413-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.